Skip to Content
Merck
All Photos(1)

Key Documents

664936

Sigma-Aldrich

Poly[tris(2,5-bis(hexyloxy)-1,4-phenylenevinylene)-alt-(1,3-phenylenevinylene)]

Synonym(s):

PTDPV

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C68H96O6)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

mol wt

average Mn 18,400
average Mw 30,200

fluorescence

λex 448 nm; λem 518 nm in chloroform

Application

Poly[tris(2,5-bis(hexyloxy)-1,4-phenylenevinylene)-alt-(1,3-phenylenevinylene)] (PTDPV) is a conducting polymer that can be used as a donor molecule and an organic dye (green) for use in organic solar cells, drug delivery, medical imaging, and organic light emitting diodes (OLEDs).[1][2][3][4][5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theoretical and experimental study of PTDPV optical and vibrational properties and its application in white electroluminescent blends
Renzi W, et al.
Synthetic Metals, 251(33), 49-56 (2019)
Multicompartmental microcylinders
Bhaskar S, et al.
Angewandte Chemie (International Edition in English), 48(25), 4589-4593 (2009)
Engineering, characterization and directional self-assembly of anisotropically modified nanocolloids
Bhaskar S, et al.
Small, 7(6), 812-819 (2011)
Origin of the open-circuit voltage of organic thin-film solar cells based on conjugated polymers
Yamanari T, et al.
Solar Energy Materials and Solar Cells, 93(6-7), 759-761 (2009)
Spatial Analysis of Metal-PLGA Hybrid Microstructures Using 3D SERS Imaging
Strozyk MS, et al.
Advances in Functional Materials, 27(33), 1701626-1701626 (2017)

Articles

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

Materials Design Concepts for Efficient Blue OLEDs: A Joint Theoretical and Experimental Study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service