Skip to Content
Merck
All Photos(1)

Key Documents

658367

Sigma-Aldrich

3,5-Dibromo-4-methylaniline

97%

Synonym(s):

4-Amino-2,6-dibromotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€141.00

€141.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€141.00

About This Item

Empirical Formula (Hill Notation):
C7H7Br2N
CAS Number:
Molecular Weight:
264.95
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€141.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

90-94 °C

functional group

bromo

SMILES string

Cc1c(Br)cc(N)cc1Br

InChI

1S/C7H7Br2N/c1-4-6(8)2-5(10)3-7(4)9/h2-3H,10H2,1H3

InChI key

AQZDIKCNODUMNY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service