Skip to Content
Merck
All Photos(1)

Documents

657174

Sigma-Aldrich

1,6-Bis(p-acetoxycarbonylphenoxy)hexane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26O8
CAS Number:
Molecular Weight:
442.46
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

composition

dimer, <20%

SMILES string

CC(=O)OC(=O)c1ccc(OCCCCCCOc2ccc(cc2)C(=O)OC(C)=O)cc1

InChI

1S/C24H26O8/c1-17(25)31-23(27)19-7-11-21(12-8-19)29-15-5-3-4-6-16-30-22-13-9-20(10-14-22)24(28)32-18(2)26/h7-14H,3-6,15-16H2,1-2H3

InChI key

UPRXYEYFWUUHHL-UHFFFAOYSA-N

Application

Monomer precursor for the synthesis of biodegradable polymers with drug delivery applications.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matt J Kipper et al.
Biomaterials, 23(22), 4405-4412 (2002-09-11)
The fabrication, morphological characterization, and drug release kinetics from microspheres of three bioerodible polyanhydrides, poly[1,6-bis(p-carboxyphenoxy)hexane] (poly(CPH)), poly(sebacic anhydride) (poly(SA)), and the copolymer poly(CPH-co-SA) 50:50 (CPH:SA 50:50) is reported. The fabrication technique yields microspheres with different morphologies for each of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service