Skip to Content
Merck
All Photos(7)

Key Documents

656453

Sigma-Aldrich

1,3,5-Tris(4-iodophenyl)benzene

greener alternative

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H15I3
CAS Number:
Molecular Weight:
684.09
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
To inquire about this product 656453, please contact your local Merck office or dealer. Contact Technical Service

Assay

98%

form

solid

greener alternative product score

old score: 100
new score: 16
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Design for Energy Efficiency
Use of Renewable Feedstocks
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

254-260 °C (lit.)

greener alternative category

SMILES string

Ic1ccc(cc1)-c2cc(cc(c2)-c3ccc(I)cc3)-c4ccc(I)cc4

InChI

1S/C24H15I3/c25-22-7-1-16(2-8-22)19-13-20(17-3-9-23(26)10-4-17)15-21(14-19)18-5-11-24(27)12-6-18/h1-15H

InChI key

KGLWDSJGGFTHHD-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Atom Economy”, “Enhanced Energy Efficiency”, “Use of Renewable Feedstock” and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Application

Building block in a synthesis of C3-symmetric nano-sized polyaromatics[1] and molecular propellers.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

European Journal of Organic Chemistry, 4003-4003 (2004)
Christopher D Simpson et al.
Journal of the American Chemical Society, 126(10), 3139-3147 (2004-03-12)
In a polymer analogous approach, large dendritic oligophenylenes containing benzene and tetraphenylmethane cores are transformed via oxidative cyclodehydrogenation to novel propeller-shaped molecules with large polycyclic aromatic hydrocarbon units as "blades". Structure analysis is performed by a combination of MALDI-TOF mass

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service