Skip to Content
Merck
All Photos(1)

Key Documents

653713

Sigma-Aldrich

4-Bromo-2-chloro-6-(trifluoromethyl)aniline

95%

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H4BrClF3N
CAS Number:
Molecular Weight:
262.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.545 (lit.)

density

1.769 g/mL at 25 °C (lit.)

SMILES string

Nc1c(Cl)cc(Br)cc1C(F)(F)F

InChI

1S/C7H4BrClF3N/c8-3-1-4(7(10,11)12)6(13)5(9)2-3/h1-2H,13H2

InChI key

GAOGWONUOUYZFD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service