Skip to Content
Merck
All Photos(1)

Key Documents

653012

Sigma-Aldrich

1-(2-Chloro-6-fluorobenzyl)piperazine

puriss., 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14ClFN2
CAS Number:
Molecular Weight:
228.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

puriss.

Assay

98%

refractive index

n20/D 1.5470 (lit.)
n20/D 1.5470

density

1.202 g/mL at 25 °C (lit.)
1.2042 g/mL at 25 °C

SMILES string

Fc1cccc(Cl)c1CN2CCNCC2

InChI

1S/C11H14ClFN2/c12-10-2-1-3-11(13)9(10)8-15-6-4-14-5-7-15/h1-3,14H,4-8H2

InChI key

PBJVZHBDDDMHIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service