Skip to Content
Merck
All Photos(1)

Key Documents

652032

Sigma-Aldrich

2,4,5-Trichloropyrimidine

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€57.00
5 G
€137.00

€57.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€57.00
5 G
€137.00

About This Item

Empirical Formula (Hill Notation):
C4HCl3N2
CAS Number:
Molecular Weight:
183.42
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€57.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

99%

refractive index

n20/D 1.574 (lit.)

density

1.6001 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Clc1cnc(Cl)nc1Cl

InChI

1S/C4HCl3N2/c5-2-1-8-4(7)9-3(2)6/h1H

InChI key

GIKMWFAAEIACRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>229.8 °F

Flash Point(C)

> 109.9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suman Rao et al.
Cell chemical biology, 26(6), 818-829 (2019-04-16)
Covalent kinase inhibitors, which typically target cysteine residues, represent an important class of clinically relevant compounds. Approximately 215 kinases are known to have potentially targetable cysteines distributed across 18 spatially distinct locations proximal to the ATP-binding pocket. However, only 40

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service