Skip to Content
Merck
All Photos(3)

Documents

649546

Sigma-Aldrich

2,6-Bis(bromomethyl)naphthalene

95%

Synonym(s):

2,6-Di(bromomethyl)naphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10Br2
CAS Number:
Molecular Weight:
314.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

183-187 °C (lit.)

SMILES string

BrCc1ccc2cc(CBr)ccc2c1

InChI

1S/C12H10Br2/c13-7-9-1-3-11-6-10(8-14)2-4-12(11)5-9/h1-6H,7-8H2

InChI key

JRHSGFPVPTWMND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service