Skip to Content
Merck
All Photos(1)

Key Documents

646180

Sigma-Aldrich

5-(5-Bromo-2-thienyl)-1H-tetrazole solution

30% in DMF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BrN4S
CAS Number:
Molecular Weight:
231.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

concentration

30% in DMF

refractive index

n20/D 1.4640

bp

157-158 °C

density

1.087 g/mL at 25 °C

SMILES string

Brc1ccc(s1)-c2nnn[nH]2

InChI

1S/C5H3BrN4S/c6-4-2-1-3(11-4)5-7-9-10-8-5/h1-2H,(H,7,8,9,10)

InChI key

GJHHXCMGAACACG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Solid Material is a potential explosive (DSC on solid 806 J/g at 239°C).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.9 °F - closed cup

Flash Point(C)

54.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service