Skip to Content
Merck
All Photos(2)

Key Documents

642339

Sigma-Aldrich

IR-797 chloride

Dye content 70 %

Synonym(s):

2-[2-[2-Chloro-3-[2-(1,3-dihydro-1,3,3-trimethyl-2H-indol-2-ylidene)-ethylidene]-1-cyclopenten-1-yl]-ethenyl]-1,3,3-trimethyl-3H-indolium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€273.00

€273.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€273.00

About This Item

Empirical Formula (Hill Notation):
C31H34Cl2N2
CAS Number:
Molecular Weight:
505.52
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€273.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

Quality Level

composition

Dye content, 70%

mp

116.7-128.0 °C (lit.)

λmax

797 nm in methanol

SMILES string

[Cl-].CN1c2ccccc2C(C)(C)\C1=C\C=C3/CCC(\C=C\C4=[N+](C)c5ccccc5C4(C)C)=C3Cl

InChI

1S/C31H34ClN2.ClH/c1-30(2)23-11-7-9-13-25(23)33(5)27(30)19-17-21-15-16-22(29(21)32)18-20-28-31(3,4)24-12-8-10-14-26(24)34(28)6;/h7-14,17-20H,15-16H2,1-6H3;1H/q+1;/p-1

InChI key

OATKVFFCVUJOEX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Solvent soluble, laser and near-IR dye

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service