Skip to Content
Merck
All Photos(1)

Key Documents

642088

Sigma-Aldrich

4-(Trifluoromethyl)phenethyl alcohol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€81.40

€81.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€81.40

About This Item

Linear Formula:
CF3C6H4CH2CH2OH
CAS Number:
Molecular Weight:
190.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€81.40


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.463 (lit.)

density

1.253 g/mL at 25 °C (lit.)

functional group

fluoro
hydroxyl

SMILES string

OCCc1ccc(cc1)C(F)(F)F

InChI

1S/C9H9F3O/c10-9(11,12)8-3-1-7(2-4-8)5-6-13/h1-4,13H,5-6H2

InChI key

SXMYWTQEZRZKBK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service