Skip to Content
Merck
All Photos(1)

Key Documents

641928

Sigma-Aldrich

Ytterbium(III) oxide

nanoparticles, <80 nm (BET), ≥99.9% trace metals basis, 5 wt % dispersion in H2O

Synonym(s):

Diytterbium trioxide, Ytterbia, Ytterbium trioxide 3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Yb2O3
CAS Number:
Molecular Weight:
394.08
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99.9% trace metals basis

form

nanoparticles

reaction suitability

reagent type: catalyst
core: ytterbium

concentration

5 wt % dispersion in H2O

particle size

<40 nm (XRD)
<80 nm (BET)

SMILES string

O=[Yb]O[Yb]=O

InChI

1S/3O.2Yb

InChI key

FIXNOXLJNSSSLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengze Yu et al.
ACS nano, 9(11), 11064-11074 (2015-10-13)
Photodynamic therapy (PDT) is a well-established modality for cancer therapy, which locally kills cancer cells when light irradiates a photosensitizer. However, conventional PDT is often limited by the extremely short lifespan and severely limited diffusion distance of reactive oxygen species

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service