640522
Diethyl 3-butenylphosphonate
95%
Synonym(s):
P-3-Buten-1-yl-phosphonic acid diethyl ester
About This Item
Recommended Products
Assay
95%
reaction suitability
reaction type: C-C Bond Formation
bp
55 °C/0.55 mmHg (lit.)
density
1.005 g/mL at 25 °C (lit.)
SMILES string
CCOP(=O)(CCC=C)OCC
InChI
1S/C8H17O3P/c1-4-7-8-12(9,10-5-2)11-6-3/h4H,1,5-8H2,2-3H3
InChI key
NUDGAPYPOAQVLP-UHFFFAOYSA-N
Application
- Bicyclo[3.3.1]alkenone framework compounds by gold-catalyzed Diels-Alder reactions
- Antiviral C-5-substituted pyrimidine acyclic nucleoside phosphonates selected as human thymidylate kinase substrates
- Antimalarials via halogenation of di-Et butenylphosphonate
- Bisphosphonate enynes
Reactant for:
- Remote supramolecular control of catalyst selectivity in hydroformlyation of alkenes
- Intermolecular metal-catalyzed carbenoid cyclopropanations
- Chemoenzymatic synthesis of phosphic derivatives of carbohydrates
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
222.1 °F - closed cup
Flash Point(C)
105.6 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service