Skip to Content
Merck
All Photos(1)

Documents

639850

Sigma-Aldrich

2-Bromo-1-methyl-1H-imidazole

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5BrN2
CAS Number:
Molecular Weight:
161.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

refractive index

n20/D 1.5440 (lit.)

bp

172 °C (lit.)

density

1.649 g/mL at 25 °C (lit.)

SMILES string

Cn1ccnc1Br

InChI

1S/C4H5BrN2/c1-7-3-2-6-4(7)5/h2-3H,1H3

InChI key

BANOTGHIHYMTDL-UHFFFAOYSA-N

Application

2-Bromo-1-methyl-1H-imidazole can be used in the synthesis of the following:
  • N-(4-methoxyphenyl)-1-methyl-1H-imidazol-2-amine via C-N coupling with p-anisidine
  • 1,2-bis[2-methyl-5-(1-methyl-1H-imidazol-2-yl)-3-thienyl]-cyclopentene, a ligand which can form bimetallic platinum(II) complex with potent cytotoxic activity

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"Non-Switching 1, 2-Dithienylethene-based Diplatinum (II) Complex Showing High Cytotoxicity"
Presa A, et al.
Inorganic Chemistry, 55(11), 5356-5364 (2016)
"CN Bond forming reactions in the synthesis of substituted 2-aminoimidazole derivatives"
SanJuan-Gomez A, et al.
ARKIVOC (Gainesville, FL, United States), 2, 44-56 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service