Skip to Content
Merck
All Photos(1)

Key Documents

639257

Sigma-Aldrich

Iodomethane-13C,d2

≥98 atom % 13C, ≥98 atom % D, ≥99% (CP), contains copper as stabilizer

Synonym(s):

Methyl Iodide-13C,d2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€447.00
5 G
€2,020.00

€447.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
€447.00
5 G
€2,020.00

About This Item

Linear Formula:
H13CD2I
CAS Number:
Molecular Weight:
144.94
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€447.00


Please contact Customer Service for Availability

Request more information

isotopic purity

≥98 atom % 13C
≥98 atom % D

Quality Level

Assay

≥99% (CP)

contains

copper as stabilizer

bp

42 °C (lit.)

mp

-66.5 °C

density

2.327 g/mL at 25 °C

application(s)

agriculture
environmental

mass shift

M+3

storage temp.

2-8°C

SMILES string

[2H][13CH]([2H])I

InChI

1S/CH3I/c1-2/h1H3/i1+1D2

InChI key

INQOMBQAUSQDDS-WGVGGRBOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service