Skip to Content
Merck
All Photos(1)

Key Documents

638757

Sigma-Aldrich

3-Fluoro-4-pyridinecarboxylic acid

97%

Synonym(s):

3-Fluoroisonicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4FNO2
CAS Number:
Molecular Weight:
141.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)c1ccncc1F

InChI

1S/C6H4FNO2/c7-5-3-8-2-1-4(5)6(9)10/h1-3H,(H,9,10)

InChI key

POLXLLIQWDBJMD-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Sutovska et al.
General physiology and biophysics, 34(2), 167-176 (2015-03-03)
Previously, therapeutic potency of CRAC channels blocker was evidenced as a significant decrease in airway smooth muscle hyperreactivity, antitussive and anti-inflammatory effects. The major role of the respiratory epithelium in asthma pathogenesis was highlighted only recently and CRAC channels were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service