Skip to Content
Merck
All Photos(1)

Key Documents

638013

Sigma-Aldrich

4-Diphenylphosphanylbenzoic acid, 2-(trimethylsilyl)ethyl ester solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27O2PSi
CAS Number:
Molecular Weight:
406.53
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

concentration

0.5 M in THF

density

0.925 g/mL at 25 °C

functional group

phosphine

SMILES string

C[Si](C)(C)CCOC(=O)c1ccc(cc1)P(c2ccccc2)c3ccccc3

InChI

1S/C24H27O2PSi/c1-28(2,3)19-18-26-24(25)20-14-16-23(17-15-20)27(21-10-6-4-7-11-21)22-12-8-5-9-13-22/h4-17H,18-19H2,1-3H3

InChI key

QTYUNPJLGFKKRV-UHFFFAOYSA-N

Application

4-Diphenylphosphanyl-benzoic acid 2-trimethylsilanyl-ethyl ester (DPPBE) was developed as an efficient alternative to triphenylphosphine in the Mitsunobu reaction since the phosphine oxide formed during the reaction can be easily removed during workup.[1]
Reagent for:
  • Synthesis of alkenes
  • Methylation of aldehydes
  • Mitsunobu reactions

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removable phosphine reagents for the Mitsunobu reaction.
Yoakim C, et al.
Synlett, 2003(04), 0473-0476 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service