All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CH3BF3K
CAS Number:
Molecular Weight:
121.94
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
form
solid
impurities
≤2 mol water
mp
168-183 °C
SMILES string
[K+].C[B-](F)(F)F
InChI
1S/CH3BF3.K/c1-2(3,4)5;/h1H3;/q-1;+1
InChI key
XMQFVHVGFQJMFF-UHFFFAOYSA-N
Application
Reactant for:
- Preparation of modified purines via palladium-catalyzed cross-coupling reactions with halopurines
- Suzuki-Miyaura cross-coupling reactions with aryl chlorides
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service