637866
Aluminum cerium oxide
nanopowder, ≤80 nm particle size (BET), 99% trace metals basis
Synonym(s):
Cerium aluminate, Cerium aluminum oxide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
99% trace metals basis
form
nanopowder
reaction suitability
reagent type: catalyst
core: cerium
particle size
≤80 nm (BET)
SMILES string
[Ce+3].[O-][Al]([O-])[O-]
InChI
1S/Al.Ce.3O/q;+3;3*-1
InChI key
GFIRVRHWPCRRBW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Product of Engi-Mat Co.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Three approaches generate white light, including LED-based down-conversion for broader applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service