Skip to Content
Merck
All Photos(1)

Key Documents

636258

Sigma-Aldrich

Isoxazole-5-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3NO3
CAS Number:
Molecular Weight:
113.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Assay

97%

form

solid

mp

144-148 °C (lit.)

SMILES string

OC(=O)c1ccno1

InChI

1S/C4H3NO3/c6-4(7)3-1-2-5-8-3/h1-2H,(H,6,7)

InChI key

MIIQJAUWHSUTIT-UHFFFAOYSA-N

General description

Isoxazole-5-carboxylic acid can be obtained via dehydration of 5-trichloromethylisoxazole.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Efficient synthesis and dehydration reaction of trichloromethylated 2-(3-phenyl-5-hydroxy-4, 5-dihydro-1H-pyrazol-1-yl)-4-aryl-5-alkylthiazoles"
Bonacorso.GH, et al.
Heteroatom Chem., 14(02), 132-137 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service