Skip to Content
Merck
All Photos(1)

Key Documents

634794

Sigma-Aldrich

(S)-(−)-1-Boc-3-aminopyrrolidine

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€45.80
5 G
€135.00

€45.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€45.80
5 G
€135.00

About This Item

Empirical Formula (Hill Notation):
C9H18N2O2
CAS Number:
Molecular Weight:
186.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€45.80


Please contact Customer Service for Availability

Request a Bulk Order

Assay

95%

optical activity

[α]20/D −2°, c = 1 in chloroform

refractive index

n20/D 1.4720 (lit.)

bp

216-217 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N1CC[C@H](N)C1

InChI

1S/C9H18N2O2/c1-9(2,3)13-8(12)11-5-4-7(10)6-11/h7H,4-6,10H2,1-3H3/t7-/m0/s1

InChI key

CMIBWIAICVBURI-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(-)-1-Boc-3-aminopyrrolidine may be used to prepare:
  • Descarboxamide analog of 4-N-(Nω-nitro-L-argininyl)-trans-4-amino-L-proline amide.[1]
  • Histamine 3 (H3) receptor antagonists containing pyrrolidin-3-yl-N-methylbenzamide moiety.[2]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective l-nitroargininylaminopyrrolidine and L-nitroargininylaminopiperidine neuronal nitric oxide synthase inhibitors.
Seo J, et al.
Bioorganic & Medicinal Chemistry, 15(5), 1928-1938 (2007)
Pyrrolidin-3-yl-N-methylbenzamides as potent histamine 3 receptor antagonists.
Zhou D, et al.
Bioorganic & Medicinal Chemistry Letters, 21(19), 5957-5960 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service