Skip to Content
Merck
All Photos(1)

Key Documents

633267

Sigma-Aldrich

Dicyclohexylcarbodiimide solution

60 wt. % in xylenes

Synonym(s):

N,N′-Methanetetraylbis[cyclohexanamine], DCC, NSC 30022, NSC 53373, NSC 57182

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22N2
CAS Number:
Molecular Weight:
206.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Coupling Reactions

concentration

60 wt. % in xylenes

refractive index

n20/D 1.503

density

0.898 g/mL at 25 °C

SMILES string

C1CCC(CC1)N=C=NC2CCCCC2

InChI

1S/C13H22N2/c1-3-7-12(8-4-1)14-11-15-13-9-5-2-6-10-13/h12-13H,1-10H2

InChI key

QOSSAOTZNIDXMA-UHFFFAOYSA-N

Application

Catalyst for Mitsunobu reaction to synthesize tetramethylpyrazine derivatives

Coupling agent for generation of graphene and graphene oxide sheets supported on silica

Endophytic bacterium L14 biomass inhibitor

Used to produce:
  • Multifunctional aqueous nanocrystals stabilized by hyperbranched polyglycerol
  • MnO2 nanosheets attached to Au nanoparticles on carbon nantubes
  • Functionalized single-walled carbon nantotube arrays

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F

Flash Point(C)

30 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siobhan P Brown et al.
American heart journal, 169(3), 334-341 (2015-03-03)
The Resuscitation Outcomes Consortium is conducting a randomized trial comparing survival with hospital discharge after continuous chest compressions without interruption for ventilation versus currently recommended American Heart Association cardiopulmonary resuscitation with interrupted chest compressions in adult patients with out-of-hospital cardiac
Marc Aprahamian et al.
Chembiochem : a European journal of chemical biology, 12(5), 777-783 (2011-03-04)
Myo-inositol trispyrophosphate (ITPP), a synthetic allosteric effector of hemoglobin, increases the regulated oxygen-releasing capacity of red blood cells (RBCs), leading to suppression of hypoxia-inducible factor 1α (HIF-1α) and to down-regulation of hypoxia-inducible genes such as vascular endothelial growth factor (VEGF).
Andreia Biolo et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(6), 1926-1929 (2009-02-11)
A major determinant of maximal exercise capacity is the delivery of oxygen to exercising muscles. myo-Inositol trispyrophosphate (ITPP) is a recently identified membrane-permeant molecule that causes allosteric regulation of Hb oxygen binding affinity. In normal mice, i.p. administration of ITPP
Kenji Mizutani et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(33), 13474-13479 (2011-08-05)
The prokaryotic V-ATPase of Enterococcus hirae, closely related to the eukaryotic enzymes, provides a unique opportunity to study the ion-translocation mechanism because it transports Na(+), which can be detected by radioisotope (22Na(+)) experiments and X-ray crystallography. In this study, we
Marten Staal et al.
Plant physiology, 155(4), 2049-2055 (2011-02-02)
In Arabidopsis (Arabidopsis thaliana; Columbia-0) roots, the so-called zone of cell elongation comprises two clearly different domains: the transition zone, a postmeristematic region (approximately 200-450 μm proximal of the root tip) with a low rate of elongation, and a fast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service