Skip to Content
Merck
All Photos(1)

Key Documents

632406

Sigma-Aldrich

Indole-6-carboxaldehyde

97%

Synonym(s):

6-Formylindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

127-131 °C (lit.)

SMILES string

O=Cc1ccc2cc[nH]c2c1

InChI

1S/C9H7NO/c11-6-7-1-2-8-3-4-10-9(8)5-7/h1-6,10H

InChI key

VSPBWOAEHQDXRD-UHFFFAOYSA-N

Application

  • Reactant in preparation of analogs of botulinum neurotoxin serotype A protease inhibitors
  • Reactant in synthesis of stilbene-based antitumor agents
  • Reactant in acid-catalyzed preparation of aromatic gem-dihalides from aldehydes and acid halides

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niina Tani et al.
Bioorganic & medicinal chemistry, 22(23), 6655-6664 (2014-12-03)
Inhibition of CYP2A6-mediated nicotine metabolism can reduce cigarette smoking. We sought potent and selective CYP2A6 inhibitors to be used as leads for drugs useful in smoking reduction therapy, by evaluating CYP2A6 inhibitory effect of novel formyl, alkyl amine or carbonitrile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service