Skip to Content
Merck
All Photos(1)

Documents

615897

Sigma-Aldrich

Phenethyl-1,1,2,2-d4-amine

98 atom % D, 98% (CP)

Synonym(s):

2-Phenylethyl-1,1,2,2-d4 amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CD2CD2NH2
Molecular Weight:
125.20
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

bp

197-200 °C (lit.)

density

0.993 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H]C([2H])(N)C([2H])([2H])c1ccccc1

InChI

1S/C8H11N/c9-7-6-8-4-2-1-3-5-8/h1-5H,6-7,9H2/i6D2,7D2

InChI key

BHHGXPLMPWCGHP-KXGHAPEVSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service