Skip to Content
Merck
All Photos(1)

Documents

614769

Sigma-Aldrich

1-Bromopropane-3,3,3-d3

99 atom % D

Synonym(s):

Propyl bromide-3,3,3-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CH2CH2Br
CAS Number:
Molecular Weight:
126.01
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

Quality Level

bp

71 °C (lit.)

mp

-110 °C (lit.)

density

1.387 g/mL at 25 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCBr

InChI

1S/C3H7Br/c1-2-3-4/h2-3H2,1H3/i1D3

InChI key

CYNYIHKIEHGYOZ-FIBGUPNXSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Ozone 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Liver,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service