Skip to Content
Merck
All Photos(1)

Documents

614505

Sigma-Aldrich

1-Bromononane-1,1,2,2-d4

98 atom % D

Synonym(s):

n-Nonyl bromide-1,1,2,2-d4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CD2CD2Br
Molecular Weight:
211.18
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

bp

201 °C (lit.)

density

1.11 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H]C([2H])(Br)C([2H])([2H])CCCCCCC

InChI

1S/C9H19Br/c1-2-3-4-5-6-7-8-9-10/h2-9H2,1H3/i8D2,9D2

InChI key

AYMUQTNXKPEMLM-LZMSFWOYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F

Flash Point(C)

90.00 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service