Skip to Content
Merck
All Photos(1)

Key Documents

609706

Sigma-Aldrich

Carbon monoxide-18O

95 atom % 18O, 99% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€877.00

€877.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 L
€877.00

About This Item

Linear Formula:
C18O
CAS Number:
Molecular Weight:
30.01
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12

€877.00


Please contact Customer Service for Availability

Request more information

isotopic purity

95 atom % 18O

Quality Level

Assay

99% (CP)

mass shift

M+2

SMILES string

[C-]#[18O+]

InChI

1S/CO/c1-2/i2+2

InChI key

UGFAIRIUMAVXCW-HQMMCQRPSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard 1 L quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 20 psig for 1 L. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951 or Z187224
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Gas 1A - Press. Gas Compr. Gas - Repr. 1A - STOT RE 1 Inhalation

Target Organs

Heart

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service