Skip to Content
Merck
All Photos(1)

Documents

608890

Sigma-Aldrich

Phthalimide-15N

98 atom % 15N

Synonym(s):

1,3-Dihydro-1,3-dioxoisoindole-15N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H515NO2
Molecular Weight:
148.12
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % 15N

form

solid

mp

232-235 °C (lit.)

mass shift

M+1

SMILES string

O=C1[15NH]C(=O)c2ccccc12

InChI

1S/C8H5NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-4H,(H,9,10,11)/i9+1

InChI key

XKJCHHZQLQNZHY-QBZHADDCSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F

Flash Point(C)

150 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service