Skip to Content
Merck
All Photos(1)

Documents

607223

Sigma-Aldrich

Carbon-13C dioxide-18O2

99 atom % 13C, 97 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C18O2
CAS Number:
Molecular Weight:
49.00
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:

isotopic purity

99 atom % 13C
97 atom % 18O

form

gas

mass shift

M+5

SMILES string

[18O]=[13C]=[18O]

InChI

1S/CO2/c2-1-3/i1+1,2+2,3+2

InChI key

CURLTUGMZLYLDI-UKEFCNGDSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service