Skip to Content
Merck
All Photos(1)

Key Documents

60713

Sigma-Aldrich

Ketene diethyl acetal

purum, ≥95.0% (GC)

Synonym(s):

1,1-Diethoxyethene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2
CAS Number:
Molecular Weight:
116.16
Beilstein:
471200
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥95.0% (GC)

refractive index

n20/D 1.414

bp

49-50 °C (lit.)

density

0.880 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=C)OCC

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service