Skip to Content
Merck
All Photos(1)

Documents

59750

Sigma-Aldrich

Isopropyltriphenylphosphonium bromide – sodium amide mixture

1 g contains ~2.3 mmol phosphonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

quality

1 g contains ~2.3 mmol phosphonium bromide

storage temp.

2-8°C

SMILES string

[Br-].N[Na].CC(C)[P+](c1ccccc1)(c2ccccc2)c3ccccc3

Quantity

1 mL contains ~1.6 mmol phosphonium bromide

Other Notes

Ready-to-use mixture for the preparation of the corresponding ylide in THF

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Schlosser et al.
Chimia, 36, 396-396 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service