Skip to Content
Merck
All Photos(1)

Key Documents

575801

Sigma-Aldrich

Poly(pyromellitic dianhydride-co-4,4′-oxydianiline), amic acid solution

15.0-16.0 wt. % in NMP

Synonym(s):

Pyre-M.L.® RC-5019, Polyamic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€92.90
1 L
€282.00

€92.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€92.90
1 L
€282.00

About This Item

Linear Formula:
(C12H12N2O .C10H2O6)x
UNSPSC Code:
12162002
NACRES:
NA.23

€92.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

concentration

15.0-16.0 wt. % in NMP

refractive index

n20/D 1.507

viscosity

50-70 poise

density

1.166 g/mL at 25 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

  • Synthesis of nano-polyimide for microelectronic applications: This study discusses the synthesis processes from Poly (amic acid) (PAA) of poly (pyromellitic dianhydride-co-4,4′-oxydianiline) and its applications in microelectronics, highlighting its potential in advanced fabrication techniques (Ebnalwaled et al., 2016).

Legal Information

Pyre-M.L. is a registered trademark of Industrial Summit Technologies Corp.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuela Loeblein et al.
Small (Weinheim an der Bergstrasse, Germany), 11(48), 6425-6434 (2015-10-21)
Polyimides (PIs) have been praised for their high thermal stability, high modulus of elasticity and tensile strength, ease of fabrication, and moldability. They are currently the standard choice for both substrates for flexible electronics and space shielding, as they render
Nurit Atar et al.
ACS applied materials & interfaces, 7(22), 12047-12056 (2015-05-07)
In low Earth orbit (LEO), hazards such as atomic oxygen (AO) or electrostatic discharge (ESD) degrade polymeric materials, specifically, the extensively used polyimide (PI) Kapton. We prepared PI-based nanocomposite films that show both AO durability and ESD protection by incorporating

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service