Skip to Content
Merck
All Photos(1)

Documents

575313

Sigma-Aldrich

Potassium tetraiodoplatinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I4K2Pt
CAS Number:
Molecular Weight:
780.89
UNSPSC Code:
12352302

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service