Skip to Content
Merck
All Photos(1)

Key Documents

573310

Sigma-Aldrich

(S)-(−)-Tetrahydrofuran-2-carboxamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D −75°, c = 1 in H2O

mp

82-86 °C (lit.)

SMILES string

NC(=O)[C@@H]1CCCO1

InChI

1S/C5H9NO2/c6-5(7)4-2-1-3-8-4/h4H,1-3H2,(H2,6,7)/t4-/m0/s1

InChI key

GXHAENUAJYZNOA-BYPYZUCNSA-N

Application

(S)-(−)-Tetrahydrofuran-2-carboxamide can be used as a precursor for the preparation of (S)-tetrahydrofuran-2-carbonitrile (THFCN) via dehydration reaction using thionyl chloride and DMF. THFCN can be further methylated with methylmagnesium chloride (Grignard reagent) to produce (S)-1-(tetrahydrofuran-2-yl)ethenone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved synthetical route of (S)?-?1-?(tetrahydrofuran-?2-?yl)?ethanone
Chai H-w, et al.
Jing Xi Hua Gong Zhong Jian Ti / Fine Chemical Intermediates, 45(1), 39-41 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service