Skip to Content
Merck
All Photos(1)

Key Documents

570141

Sigma-Aldrich

Tris(dimethylamino)chlorosilane

≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€389.00

€389.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€389.00

About This Item

Linear Formula:
ClSi(N(CH3)2)3
CAS Number:
Molecular Weight:
195.77
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€389.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

bp

78 °C/14 mmHg (lit.)

density

0.973 g/mL at 25 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CN(C)[Si](Cl)(N(C)C)N(C)C

InChI

1S/C6H18ClN3Si/c1-8(2)11(7,9(3)4)10(5)6/h1-6H3

InChI key

YLZCZVGQEADVNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.0 °F - closed cup

Flash Point(C)

58.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service