Skip to Content
Merck
All Photos(2)

Key Documents

569801

Sigma-Aldrich

Pyridinium chlorochromate on silica gel

200-400 mm bead size, ~20 % (silica gel), on silica gel

Synonym(s):

PCC on silica gel

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€269.00
100 G
€375.00

€269.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€269.00
100 G
€375.00

About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

€269.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

reaction suitability

reagent type: oxidant

bead size

200-400 mm

capacity

~20 % (silica gel)

storage temp.

2-8°C

SMILES string

O[Cr](Cl)(=O)=O.c1ccncc1

InChI

1S/C5H5N.ClH.Cr.H2O.2O/c1-2-4-6-5-3-1;;;;;/h1-5H;1H;;1H2;;/q;;+2;;;/p-2

InChI key

JWIPGAFCGUDKEY-UHFFFAOYSA-L

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Ox. Sol. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Functionalized silica gel is used in size-exclusion chromatography of cationic polymers and in the separation of sugars, steroids, cholesterol, and triglycerides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service