Skip to Content
Merck
All Photos(1)

Key Documents

567221

Sigma-Aldrich

Tris(3,3′,3″-phosphinidynetris(benzenesulfonato)palladium(0) nonasodium salt nonahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€281.00

€281.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€281.00

About This Item

Linear Formula:
Pd(P(C6H4SO3·Na)3)3 · 9H2O
CAS Number:
Molecular Weight:
1982.89
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€281.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

>300 °C (lit.)

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Pd].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-]S(=O)(=O)c1cccc(c1)P(c2cccc(c2)S([O-])(=O)=O)c3cccc(c3)S([O-])(=O)=O.[O-]S(=O)(=O)c4cccc(c4)P(c5cccc(c5)S([O-])(=O)=O)c6cccc(c6)S([O-])(=O)=O.[O-]S(=O)(=O)c7cccc(c7)P(c8cccc(c8)S([O-])(=O)=O)c9cccc(c9)S([O-])(=O)=O

InChI

1S/3C18H15O9PS3.9Na.9H2O.Pd/c3*19-29(20,21)16-7-1-4-13(10-16)28(14-5-2-8-17(11-14)30(22,23)24)15-6-3-9-18(12-15)31(25,26)27;;;;;;;;;;;;;;;;;;;/h3*1-12H,(H,19,20,21)(H,22,23,24)(H,25,26,27);;;;;;;;;;9*1H2;/q;;;9*+1;;;;;;;;;;/p-9

InChI key

RZJLWYKBARRAJM-UHFFFAOYSA-E

Application

Tris(3,3′,3″-phosphinidynetris(benzenesulfonato)palladium(0) nonasodium salt nonahydrate (Pd-TPPTS complex) supported by layered double hydroxide (LDH) can be used as an efficient catalyst in the cycloisomerization of acetylenic carboxylic acids to the corresponding 5-membered heterocycles.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Pd heterogeneous catalysts for cycloisomerisation of acetylenic carboxylic acids
Neactu F, et al.
Green Chemistry, 12(12), 2145-2149 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service