Skip to Content
Merck
All Photos(2)

Key Documents

565814

Sigma-Aldrich

4-(N-Boc-amino)phenylboronic acid

≥95.0%

Synonym(s):

4-(Boc-amino)phenylboronic acid, 4-[(tert-Butoxycarbonyl)amino]benzeneboronic acid, C-(1,1-Dimethylethyl) N-(4-boronophenyl)carbamate, [4-(N-Bocamino)phenyl]boronic acid, [4-(tert-Butoxycarbonylamino)phenyl]boronic acid, [4-[[[(1,1-Dimethylethyl)oxy]carbonyl]amino]phenyl]boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€92.40

€92.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€92.40

About This Item

Linear Formula:
(CH3)3CO2CNHC6H4B(OH)2
CAS Number:
Molecular Weight:
237.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€92.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95.0%

form

solid

mp

199-204 °C (dec.) (lit.)

functional group

amine

SMILES string

CC(C)(C)OC(=O)Nc1ccc(cc1)B(O)O

InChI

1S/C11H16BNO4/c1-11(2,3)17-10(14)13-9-6-4-8(5-7-9)12(15)16/h4-7,15-16H,1-3H3,(H,13,14)

InChI key

UBVOLHQIEQVXGM-UHFFFAOYSA-N

Application

Boronic acid used in a study of the rhodium-catalyzed desymmetrization of a meso-cyclic allylic dicarbonate via SN2′ substitution.[1]

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frederic Menard et al.
Organic letters, 8(20), 4569-4572 (2006-09-22)
An enantio-, regio-, and diastereoselective rhodium(I)-catalyzed desymmetrization of a meso-cyclic allylic dicarbonate with organoboronic acid nucleophiles is described. The rhodium(I) catalyst formed in situ from [Rh(cod)OH]2 and Xyl-P-PHOS allowed the S(N)2' allylic substitution product to be obtained with a range

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service