Skip to Content
Merck
All Photos(1)

Key Documents

564990

Sigma-Aldrich

N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine

Synonym(s):

Ishikawa’s Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€147.00

€147.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€147.00

About This Item

Linear Formula:
CF3CHFCF2N(C2H5)2
CAS Number:
Molecular Weight:
223.16
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€147.00


Please contact Customer Service for Availability

Request a Bulk Order

refractive index

n20/D 1.3460 (lit.)

bp

56-57 °C (lit.)

density

1.230 g/mL at 25 °C (lit.)

functional group

amine
fluoro

SMILES string

CCN(CC)C(F)(F)C(F)C(F)(F)F

InChI

1S/C7H11F6N/c1-3-14(4-2)7(12,13)5(8)6(9,10)11/h5H,3-4H2,1-2H3

InChI key

BNTFCVMJHBNJAR-UHFFFAOYSA-N

General description

GC analysis includes Et2NCF=CFCF3 (two isomers), both of which are active fluorinating agents.

Application

Demonstrates utility in fluorination.[1]
Displays the ability to insert a fluoro(trifluoromethyl) methylene moiety in unsaturated alcohols.[2]
Reactant for:
  • Friedel-Crafts type arylation of hydrofluorocarbons
  • Microbial hydroxylation of antibiotics
  • Electrochemical fluorination of trialkylamines and tetraalkylammonium salts
  • Fluorination of pyrethroides

Reactant for synthesis of:
  • γ-Hydroxy-α-fluoro-α-trifluoromethylcarboxamide
  • Florfenicol and thiamphenicol

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ogu, K-i et al
Tetrahedron Letters, 39, 305-305 (1998)
Koch, G. et al
Synlett, 693-693 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service