559059
Ethyl (4-trifluoromethylbenzoyl)acetate
Synonym(s):
3-Oxo-3-(4-trifluoromethylphenyl)propanoic acid ethyl ester, Ethyl 2-4-(Trifluoromethyl)benzoylacetate, Ethyl 3-(4-trifluoromethylphenyl)-3-oxopropionate, Ethyl 3-oxo-3-4-(trifluoromethyl)phenylpropanoate
About This Item
Recommended Products
refractive index
n20/D 1.4880 (lit.)
bp
248-249 °C (lit.)
density
1.270 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)CC(=O)c1ccc(cc1)C(F)(F)F
InChI
1S/C12H11F3O3/c1-2-18-11(17)7-10(16)8-3-5-9(6-4-8)12(13,14)15/h3-6H,2,7H2,1H3
InChI key
HVHVSJPSNQIPEM-UHFFFAOYSA-N
Application
- In-catalyzed cycloisomerization
- Chiral Lewis base-catalyzed stereoselective reduction with trichlorosilane and water
- Guanidine-catalyzed enantioselective Michael reactions
- Transesterification reactions
- Intermolecular coupling reactions
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service