Skip to Content
Merck
All Photos(1)

Key Documents

558966

Sigma-Aldrich

1-(2-Furoyl)piperazine

97%

Synonym(s):

2-Furoic piperazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O2
CAS Number:
Molecular Weight:
180.20
Beilstein:
880196
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%
To inquire about this product 558966, please contact your local Merck office or dealer. Contact Technical Service

Assay

97%

mp

67-70 °C (lit.)

SMILES string

O=C(N1CCNCC1)c2ccco2

InChI

1S/C9H12N2O2/c12-9(8-2-1-7-13-8)11-5-3-10-4-6-11/h1-2,7,10H,3-6H2

InChI key

SADPINFEWFPMEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-(2-Furoyl)piperazine may be used to synthesize:
  • 2-[4-(2-furoyl) piperazin-1-yl]-4-amino-6-benzyloxy-7-methoxyquinazoline hydrochloride monohydrate[1]
  • 2-[4-(2-furoyl)piperazin-1-yl]-4-amino-7-benzyloxy-6-methoxyquinazoline hydrochloride monohydrate[1]
  • 4-amino-2-[4-(2-furoyl)piperazin-1-yl]-quinazoline hydrochloride[2]
  • 6,7-dimethoxy-2-[4-(2-furoyl)-piperazin-1-yl]quinazoline-4-one hydrate[2]
  • 1-[2-(3,4-dimethoxyphenyl)ethyl]-4-(2-furoyl)-piperazine hydrogen oxalate[2]
  • 1-(3,4-dichlorobenzyl)-4-(2-furoyl)-piperazine hydrogen oxalate[2]
  • 4-(9-fluorenyl)-1-(2-furoyl)piperazine[2]
  • 2-[4-(2-furoyl)piperazin-1-yl]-benzimidazole hydrochloride hydrate[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Search for the pharmacophore in prazosin for Transport-P.
Zunszain PA, et al.
Bioorganic & Medicinal Chemistry, 13(11), 3681-3689 (2005)
Synthesis and identification of the major metabolites of prazosin formed in dog and rat.
Althuis TH and Hess HJD.
Journal of Medicinal Chemistry, 20(1), 146-149 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service