Skip to Content
Merck
All Photos(1)

Key Documents

557943

Sigma-Aldrich

Ethyl 2-cyanobenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4CO2C2H5
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

63-67 °C (lit.)

functional group

ester
nitrile

SMILES string

CCOC(=O)c1ccccc1C#N

InChI

1S/C10H9NO2/c1-2-13-10(12)9-6-4-3-5-8(9)7-11/h3-6H,2H2,1H3

InChI key

RQQAJOJDAZNFPF-UHFFFAOYSA-N

General description

Reaction between phthalic acid monoamide and ethanol in pyridine in the presence of p-toluenesulfonyl chloride affords ethyl 2-cyanobenzoate.[1]

Application

Ethyl 2-cyanobenzoate may be used to synthesize 2-(aminomethyl)benzyl alcohol.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of ortho substituents on carbonyl carbon 13C NMR chemical shifts in substituted phenyl benzoates.
Nummert V, et al.
Journal of Physical Organic Chemistry, 22(12), 1155-1165 (2009)
5-Membered N-heterocyclic compounds by dimethyl carbonate chemistry.
Arico F, et al.
Green Chemistry, 14(1), 58-61 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service