Skip to Content
Merck
All Photos(1)

Key Documents

557749

Sigma-Aldrich

N-Boc-5-bromoindole

97%

Synonym(s):

tert-Butyl 5-bromoindole-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€98.80
25 G
€469.00

€98.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€98.80
25 G
€469.00

About This Item

Empirical Formula (Hill Notation):
C13H14BrNO2
CAS Number:
Molecular Weight:
296.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€98.80


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

56-57 °C (lit.)

functional group

bromo

SMILES string

CC(C)(C)OC(=O)n1ccc2cc(Br)ccc12

InChI

1S/C13H14BrNO2/c1-13(2,3)17-12(16)15-7-6-9-8-10(14)4-5-11(9)15/h4-8H,1-3H3

InChI key

PBWDRTGTQIXVBR-UHFFFAOYSA-N

General description

N-Boc-5-bromoindole can undergo Sonogashira coupling reaction with N,N-diisopropylprop-2-ynylamine to afford the corresponding propargylic diisopropylamine.[1] N-Boc-5-bromoindole is formed as an intermediate during the synthesis of 2-(5-substituted-1H-indol-3-yl)-N-hydroxyacetamide derivatives.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyuki Nakamura et al.
The Journal of organic chemistry, 70(6), 2357-2360 (2005-03-12)
[reaction: see text] Propargylic diisopropylamines containing heterocycles, which were prepared readily from heterocyclic bromides and propargyldiisopropylamine by the Sonogashira coupling reaction, underwent the allene transformation reaction in the presence of Pd(2)(dba)(3).CHCl(3) catalyst (2.5 mol %) and 1,2-bis[bis(pentafluorophenyl)phosphino]ethane (10 mol %)
Sylvain Petit et al.
ChemMedChem, 4(2), 261-275 (2008-12-05)
The lead compound 5-bromoindolyl-3-acetohydroxamic acid (10) was recently identified as a potent inhibitor of bacterial peptide deformylases (PDFs). The synthesis and associated activities of new variants were investigated at position 5 to optimize the fit at the S1' subsite and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service