Skip to Content
Merck
All Photos(1)

Key Documents

550612

Sigma-Aldrich

Ethyl 3-quinolinecarboxylate

97%

Synonym(s):

Quinoline-3-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO2
CAS Number:
Molecular Weight:
201.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

120 °C/0.4 mmHg (lit.)

mp

63-67 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1cnc2ccccc2c1

InChI

1S/C12H11NO2/c1-2-15-12(14)10-7-9-5-3-4-6-11(9)13-8-10/h3-8H,2H2,1H3

InChI key

OTTDACPMYLDVTL-UHFFFAOYSA-N

General description

Ethyl 3-quinolinecarboxylate can be prepared from 3-quinolinecarboxylic acid by treating with thionyl chloride followed by esterification with ethanol. Its photochemical reaction in various alcohols and cyclohexane show that the nature of the solvent has an impact on the product formation.[1]

Application

Ethyl 3-quinolinecarboxylate may be used as a starting material in the preparation of 5-quinolinemethanol[2] and ethyl 3-quinolineacetate.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

QUINOLINEMETHANOLS1.
Kaslow CE and Clark WR.
The Journal of Organic Chemistry, 18(1), 55-58 (1953)
Photochemical reactions of ethoxycarbonyl-substituted quinolines.
Ono I and Hata N.
Bulletin of the Chemical Society of Japan, 60(8), 2891-2897 (1987)
Biosynthesis of Penicillins. VI. N-2-Hydroxyethylamides of Some Polycyclic and Heterocyclic Acetic Acids as Precursors1.
Jones RG, et al.
Journal of the American Chemical Society, 70(9), 2843-2848 (1948)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service