Skip to Content
Merck
All Photos(1)

Key Documents

549959

Sigma-Aldrich

2-Benzylcyclohexanone

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2C6H9O
CAS Number:
Molecular Weight:
188.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥97%

Assay

≥97%

refractive index

n20/D 1.536 (lit.)

bp

103-105 °C/0.2 mmHg (lit.)

density

1.024 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCC1Cc2ccccc2

InChI

1S/C13H16O/c14-13-9-5-4-8-12(13)10-11-6-2-1-3-7-11/h1-3,6-7,12H,4-5,8-10H2

InChI key

CUYLPYBYCYQGCE-UHFFFAOYSA-N

General description

2-Benzylcyclohexanone is a 2-substituted cyclohexanone derivative that can be prepared by the catalytic hydrogenation of 2-benzalcyclohexanone.[1]

Application

2-Benzylcyclohexanone may be used in the preparation of ethyl 1-hydroxy-2-benzylcyclohexylacetate.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of carbanions. XIX. Alkylation of enolates from unsymmetrical ketones.
House HO, et al.
The Journal of Organic Chemistry, 36(16), 2361-2371 (1971)
Dialkylaminoalkyl Derivatives of 10,11-Dihydro-5H-dibenzo[a,d]cycloheptene and Related Compounds.
Villani FJ, et al.
Journal of Medicinal Chemistry, 5(2), 373-383 (1962)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service