Skip to Content
Merck
All Photos(1)

Key Documents

549045

Sigma-Aldrich

4-Chlorothioanisole

98%

Synonym(s):

4-Chlorophenyl methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SCH3
CAS Number:
Molecular Weight:
158.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.598 (lit.)

bp

168-169 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

SMILES string

CSc1ccc(Cl)cc1

InChI

1S/C7H7ClS/c1-9-7-4-2-6(8)3-5-7/h2-5H,1H3

InChI key

KIQQUVJOLVCZKG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariel Hasson et al.
Journal of drugs in dermatology : JDD, 9(12), 1507-1509 (2010-12-02)
The treatment of atrophic scars is difficult and dermal filler materials provide a simple alternative with immediate results. Esthélis® is an injectable non-animal crosslinked hyaluronic acid of Swiss origin characterized by a polydense cohesive matrix (CPM®) which produces a gel
I P Nnane et al.
Xenobiotica; the fate of foreign compounds in biological systems, 33(1), 83-91 (2003-01-10)
1. The biotransformation of ethyl methyl sulphide (EMS), 4-chlorophenyl methyl sulphide (CPMS) and diphenyl sulphide (DPS) to their corresponding sulphoxides by purified flavin-containing monooxygenase (FMO) is described. 2. Purified pig liver flavin-containing monooxygenase catalysed the sulphoxidation of EMS, CPMS and
Metabolism of 4-chlorophenyl methyl sulfide and its sulfone analog in cattle and sheep.
D D Oehler et al.
Archives of environmental contamination and toxicology, 12(2), 227-233 (1983-03-01)
Analysis of chlorophenylmethylsulfides in the urine of rats injected with chlorobenzene by high performance liquid chromatography.
M Yoshida et al.
Industrial health, 23(4), 283-287 (1985-01-01)
Ivo P Nnane et al.
Biomedical chromatography : BMC, 19(1), 87-98 (2004-10-08)
In this investigation, the measurement and identification of the S-oxidation products of three simple sulphides-ethyl methyl sulphide (EMS), 4-chlorophenyl methyl sulphide (CPMS) and diphenyl sulphide (DPS)-in rat urine were carried out and a study of the effects of phenobarbitone (PB)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service