Skip to Content
Merck
All Photos(2)

Key Documents

548987

Sigma-Aldrich

2,5-Dichlorobenzyl bromide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2Br
CAS Number:
Molecular Weight:
239.92
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

38-42 °C (lit.)

SMILES string

Clc1ccc(Cl)c(CBr)c1

InChI

1S/C7H5BrCl2/c8-4-5-3-6(9)1-2-7(5)10/h1-3H,4H2

InChI key

UUVDOPTUDWJHFK-UHFFFAOYSA-N

Application

2,5-Dichlorobenzyl bromide may be used in the synthesis of methyl 5-(2,5-dichlorobenzyloxy)-2-hydroxybenzoate and methyl 4-(2,5-dichlorobenzyloxy)-2-hydroxybenzoate via chemoselective alkylation of the hydroxyl group of the corresponding ester.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and bioevaluation of aryl hydroxamates distinguishing between NAD+ and ATP-dependent DNA ligases.
Kukshal V, et al.
MedChemComm, 3(4), 453-461 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service