Skip to Content
Merck
All Photos(1)

Documents

548502

Sigma-Aldrich

Mercury(II) benzoate hydrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(OCOC6H5)2 · xH2O
Molecular Weight:
442.82 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99%

form

powder

mp

166-167 °C (lit.)

SMILES string

O.O=C(O[Hg]OC(=O)c1ccccc1)c2ccccc2

InChI

1S/2C7H6O2.Hg.H2O/c2*8-7(9)6-4-2-1-3-5-6;;/h2*1-5H,(H,8,9);;1H2/q;;+2;/p-2

InChI key

FYZBDVURUZQBKY-UHFFFAOYSA-L

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service