Skip to Content
Merck
All Photos(2)

Key Documents

548456

Sigma-Aldrich

Indium(II) chloride

99.9% trace metals basis

Synonym(s):

Indium dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€57.40
10 G
€254.00

€57.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€57.40
10 G
€254.00

About This Item

Linear Formula:
InCl2
CAS Number:
Molecular Weight:
185.72
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

€57.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: indium

impurities

≤1500.0 ppm Trace Metal Analysis

mp

262 °C (dec.) (lit.)

SMILES string

Cl[InH]Cl

InChI

1S/2ClH.In.H/h2*1H;;/q;;+2;/p-2

InChI key

RLVOJNSMGYFATO-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service