Skip to Content
Merck
All Photos(1)

Key Documents

544485

Sigma-Aldrich

2-Bromo-4-chloroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(Cl)NH2
CAS Number:
Molecular Weight:
206.47
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

64-68 °C (lit.)

SMILES string

Nc1ccc(Cl)cc1Br

InChI

1S/C6H5BrClN/c7-5-3-4(8)1-2-6(5)9/h1-3H,9H2

InChI key

SYTBIFURTZACKR-UHFFFAOYSA-N

General description

2-Bromo-4-chloroaniline is a 2-bromo-4-substituted aniline that can be prepared from 4-chloroaniline via electrophilic bromination.[1]

Application

2-Bromo-4-chloroaniline may be used in the preparation of:
  • 2-bromo-4-chloro-N-tosylaniline[2]
  • 2-bromo-4-chlorothiophenol[3]
  • 7-chloro-4a-methyl-4,4a,9,9a-tetrahydro-1H-carbazol-2(3H)-one[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2-Aza-and 8-Chloro-2-aza-phenothiazine1.
Saggiomo AJ, et al.
The Journal of Organic Chemistry, 23(12), 1906-1909 (1958)
An intriguing effect of lithium perchlorate dispersed on silica gel in the bromination of aromatic compounds by N-bromosuccinimide.
Bagheri M, et al.
Canadian Journal of Chemistry, 83(2), 146-149 (2005)
Palladium-Catalyzed ?-Arylation of ?, ?-Unsaturated Ketones: Application to a One-Pot Synthesis of Tricyclic Indolines.
Hyde AM and Buchwald SL.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 120(1), 183?186-183?186 (2007)
Palladium-catalyzed coupling of 2-bromoanilines with vinylstannanes. A regiocontrolled synthesis of substituted indoles.
Krolski ME, et al.
The Journal of Organic Chemistry, 53(6), 1170-1176 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service